Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cited2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010828.3 | NP_034958.2 | 2 | 920 | ||
GenBank mRNA | AK131664.1 | - | 2 | 923 | ||
AK133791.1 | - | 2 | 921 | |||
BC057126.1 | - | 2 | 902 | 6415181 | ||
U86445.1 | - | 3 | 732 | |||
Y15163.1 | - | 2 | 899 |