Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010833.2 | NP_034963.2 | 4 | 411 | ||
GenBank mRNA | AK031171.1 | - | 4 | 401 | ||
AK088336.1 | - | 4 | 359 | |||
AK133819.1 | - | 4 | 415 | |||
AK145746.1 | - | 4 | 415 | |||
AK150869.1 | - | 4 | 414 | |||
AK153210.1 | - | 4 | 414 | |||
AK157701.1 | - | 4 | 366 | |||
BC003474.1 | - | 4 | 402 | |||
BC047366.1 | - | 4 | 405 | 3711212 | ||
M86390.1 | - | 3 | 191 | |||
S47577.1 | - | 4 | 396 |