Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198754.1 | NP_001185683.1 | 4 | 461 | ||
NM_001198755.1 | NP_001185684.1 | 4 | 488 | |||
NM_001198756.1 | NP_001185685.1 | 4 | 378 | |||
NM_031498.2 | NP_113686.1 | 4 | 505 | |||
GenBank mRNA | AF365870.1 | - | 3 | 269 | ||
AF493876.1 | - | 2 | 198 | |||
AK312839.1 | - | 4 | 489 | |||
BC008663.1 | - | 4 | 505 | 3866242 | ||
BM723861.1 | - | 4 | 378 | |||
DA248836.1 | - | 4 | 461 |