Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010934.4 | NP_035064.1 | 2 | 692 | ||
GenBank mRNA | AK132397.1 | - | 2 | 693 | ||
D63818.1 | - | 1 | 527 | |||
D63819.1 | - | 649 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC051420.1 | - | 585 | 6490858 | Chr. - - [-] on Build |
Set Membership: |
G-protein coupled receptor |