Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010942.3 | NP_035072.2 | 3 | 286 | ||
GenBank mRNA | AF035683.1 | - | 3 | 235 | ||
AK076451.1 | - | 3 | 726 | |||
AK078054.1 | - | 3 | 277 | |||
AK144503.1 | - | 3 | 261 | |||
AK148941.1 | - | 3 | 286 | |||
AX775765.1 | - | 3 | 286 | |||
BC008272.1 | - | 3 | 272 | 3603440 |