Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011049.4 | NP_035179.1 | 6 | 872 | ||
GenBank mRNA | AK051425.1 | - | 6 | 873 | ||
AK143518.1 | - | 6 | 900 | |||
AK145483.1 | - | 6 | 902 | |||
AK166188.1 | - | 6 | 1081 | |||
AK206546.1 | - | 2 | 105 | |||
BC011069.1 | - | 6 | 615 | 4006745 | ||
BC013663.1 | - | 6 | 931 | 4208974 | ||
X69025.1 | - | 6 | 894 |