Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005417.4 | NP_005408.1 | 5 | 779 | ||
NM_198291.2 | NP_938033.1 | 5 | 704 | |||
XM_011529013.2 | XP_011527315.1 | 6 | 1048 | |||
XM_017028024.1 | XP_016883513.1 | 6 | 1054 | |||
XM_017028025.1 | XP_016883514.1 | 5 | 935 | |||
XM_017028026.1 | XP_016883515.1 | 5 | 774 | |||
XM_017028027.1 | XP_016883516.1 | 3 | 351 | |||
GenBank mRNA | BC011566.1 | - | 3 | 369 | 4548399 | |
BC051270.1 | - | 3 | 432 | 4871614 |