Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253357.1 | NP_001240286.1 | 3 | 495 | ||
NM_005424.4 | NP_005415.1 | 3 | 578 | |||
XM_005271163.1 | XP_005271220.1 | 3 | 521 | |||
XM_006710869.1 | XP_006710932.1 | 3 | 712 | |||
XM_017002207.1 | XP_016857696.1 | 3 | 521 | |||
GenBank mRNA | AK225560.1 | - | 3 | 456 | ||
AK300399.1 | - | 3 | 385 | |||
AK300623.1 | - | 4 | 407 | |||
BC038239.1 | - | 3 | 480 | 5767075 | ||
EU826586.1 | - | 3 | 401 | |||
EU826587.1 | - | 3 | 401 | |||
EU826588.1 | - | 3 | 401 | |||
EU826590.1 | - | 3 | 401 | |||
X60957.1 | - | 3 | 437 |