Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011113.3 | NP_035243.1 | 5 | 553 | ||
GenBank mRNA | AK002580.1 | - | 5 | 624 | ||
AK131951.1 | - | 5 | 569 | |||
AK144796.1 | - | 5 | 570 | |||
AK146827.1 | - | 5 | 568 | |||
AK154141.1 | - | 5 | 570 | |||
AK155351.1 | - | 5 | 557 | |||
BC010309.1 | - | 4 | 1071 | |||
X62700.1 | - | 5 | 554 | |||
X62701.1 | - | 5 | 772 |