Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011116.1 | NP_035246.1 | 14 | 2141 | ||
GenBank mRNA | AF026124.1 | - | 14 | 2141 | ||
AK003045.1 | - | 1 | 356 | |||
AK153508.1 | - | 12 | 1830 | |||
BC068170.1 | - | 14 | 2137 | |||
BC076586.1 | - | 14 | 2205 | 30604186 |