Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011161.5 | NP_035291.4 | 2 | 248 | ||
GenBank mRNA | AF135185.1 | - | 2 | 228 | ||
AK144846.1 | - | 2 | 221 | |||
AK161190.1 | - | 2 | 136 | |||
AK161562.1 | - | 2 | 200 | |||
BC057211.1 | - | 2 | 184 | |||
BC064737.1 | - | 2 | 199 | 30112690 | ||
BC092526.1 | - | 2 | 248 | 6593321 |