Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpru | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083119.2 | NP_001076588.1 | 2 | 259 | ||
NM_011214.2 | NP_035344.2 | 2 | 259 | |||
GenBank mRNA | AK052720.1 | - | 2 | 234 | ||
AK148515.1 | - | 2 | 256 | |||
BC048694.1 | - | 2 | 215 | |||
BC080736.1 | - | 2 | 214 | |||
D88187.1 | - | 2 | 250 |