Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011136.2 | NP_035266.1 | 2 | 191 | ||
GenBank mRNA | AK088587.1 | - | 2 | 191 | ||
AK135080.1 | - | 2 | 192 | |||
BC058611.1 | - | 2 | 172 | 30077096 | ||
U43788.1 | - | 2 | 191 | |||
Z54283.1 | - | 2 | 194 |