Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011172.2 | NP_035302.2 | 9 | 1091 | ||
GenBank mRNA | AF120279.1 | - | 9 | 1048 | ||
AK144068.1 | - | 9 | 1061 | |||
BC008548.1 | - | 9 | 915 | |||
BC125327.1 | - | 9 | 894 | 40129842 | ||
U80020.1 | - | 9 | 1040 |