Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281731.1 | NP_001268660.1 | 11 | 1806 | ||
NM_147190.3 | NP_671723.1 | 10 | 1605 | |||
NR_104035.1 | 10 | 2654 | ||||
XM_005269220.2 | XP_005269277.1 | 10 | 1587 | |||
XM_006719687.3 | XP_006719750.1 | 11 | 1577 | |||
XM_011538975.2 | XP_011537277.1 | 11 | 1544 | |||
XM_017020202.1 | XP_016875691.1 | 11 | 1501 | |||
XM_017020203.1 | XP_016875692.1 | 12 | 1832 | |||
XM_017020205.1 | XP_016875694.1 | 11 | 1397 | |||
GenBank mRNA | AK026014.1 | - | 10 | 1411 | ||
AK055740.1 | - | 1 | 1408 | |||
AK058033.1 | - | 8 | 2143 | |||
BC032565.1 | - | 10 | 1422 | 5580489 | ||
BC033558.1 | - | 10 | 2441 |