Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146174.1 | NP_001139646.1 | 2 | 335 | ||
NM_011241.4 | NP_035371.4 | 3 | 383 | |||
GenBank mRNA | AK035571.1 | - | 3 | 345 | ||
AK042930.1 | - | 2 | 300 | |||
AK088768.1 | - | 2 | 257 | |||
AK129452.1 | - | 2 | 264 | |||
AK133905.1 | - | 2 | 300 | |||
AK145459.1 | - | 2 | 256 | |||
AK146670.1 | - | 3 | 343 | |||
AK159009.1 | - | 2 | 288 | |||
AK159525.1 | - | 2 | 300 | |||
AK159930.1 | - | 2 | 291 | |||
BC014855.1 | - | 2 | 291 | 3157225 | ||
BC052862.1 | - | 2 | 258 | 30063509 | ||
BC066213.1 | - | 2 | 299 | 30466595 | ||
BC071200.1 | - | 3 | 499 | 5032610 | ||
U08110.1 | - | 3 | 366 |