Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011259.1 | NP_035389.1 | 2, 3 | 155 | ||
GenBank mRNA | BC119153.1 | - | 2, 3 | 155 | 8733902 | |
D63356.1 | - | 2, 3 | 155 | |||
D63357.1 | - | 2, 3 | 104 |