Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011359.2 | NP_035489.2 | 4 | 451 | ||
GenBank mRNA | BC061137.1 | - | 4 | 446 | 30300758 | |
CJ093163.1 | - | 4 | 447 | |||
CJ237354.1 | - | 2 | 163 |