Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035804.1 | - | 10 | 1708 | ||
AK088317.1 | - | 7 | 2294 | |||
AK133759.1 | - | 11 | 1641 | |||
BC005553.1 | - | 11 | 1676 | 3586154 | ||
BC110668.1 | - | 11 | 2068 | 6391053 | ||
DQ186662.1 | - | 9 | 1566 | |||
Z49916.1 | - | 11 | 1979 |