Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sdf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014418.1 | - | 3 | 613 | ||
AK157860.1 | - | 3 | 478 | |||
AK163400.1 | - | 3 | 598 | |||
AK167113.1 | - | 3 | 527 | |||
BC005715.1 | - | 4 | 747 | |||
BC068152.1 | - | 3 | 853 | 30534982 | ||
D50461.1 | - | 3 | 856 | |||
U45977.1 | - | 3 | 761 | |||
U45978.1 | - | 3 | 629 |