Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164059.1 | NP_001157531.1 | 6, 7 | 1094 | ||
NM_011346.2 | NP_035476.1 | 7, 8 | 1202 | |||
GenBank mRNA | AK137509.1 | - | 7, 8 | 1114 | ||
AK157273.1 | - | 7, 8 | 1191 | |||
AK161754.1 | - | 7, 8 | 1194 | |||
AK163066.1 | - | 5, 6 | 659 | |||
AK170753.1 | - | 6, 7 | 1095 | |||
AK172116.1 | - | 7, 8 | 1192 | |||
BC052681.1 | - | 7, 8 | 1186 | 30072021 | ||
M25324.1 | - | 7, 8 | 1194 | |||
M36005.1 | - | 7, 8 | 1140 | |||
M36058.1 | - | 7, 8 | 1140 | |||
X14772.1 | - | 7, 8 | 1143 |