Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shcbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011369.2 | NP_035499.1 | 13 | 1754 | ||
GenBank mRNA | AF017152.1 | - | 13 | 1754 | ||
AK137496.1 | - | 12 | 2236 | |||
AK144966.1 | - | 13 | 1777 | |||
AK162943.1 | - | 13 | 1768 | |||
BC070455.1 | - | 13 | 1774 | 30606785 |