Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011443.3 | NP_035573.3 | 1 | 734 | ||
GenBank mRNA | AB108673.1 | - | 1 | 356 | ||
AB221649.1 | - | 1 | 323 | |||
AK131933.1 | - | 1 | 172 | |||
AK149358.1 | - | 1 | 57 | |||
BC057574.1 | - | 1 | 605 | 6413283 | ||
U31967.1 | - | 1 | 684 |