Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011495.2 | NP_035625.2 | 3 | 491 | ||
NM_173169.2 | NP_775261.2 | 3 | 491 | |||
GenBank mRNA | AK006827.1 | - | 3 | 397 | ||
AK045082.1 | - | 3 | 415 | |||
AK137080.1 | - | 3 | 403 | |||
AK137471.1 | - | 3 | 402 | |||
AK164465.1 | - | 3 | 401 | |||
BC026785.1 | - | 3 | 401 | 5150173 | ||
BC057940.1 | - | 3 | 388 | 5323295 | ||
L29479.1 | - | 3 | 393 | |||
L29480.1 | - | 3 | 393 |