Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011505.2 | NP_035635.1 | 4 | 316 | ||
GenBank mRNA | AF152924.1 | - | 4 | 306 | ||
AK012293.1 | - | 4 | 274 | |||
AK049001.1 | - | 4 | 285 | |||
BC032881.1 | - | 4 | 270 | 1224878 | ||
BY720472.1 | - | 4 | 317 |