Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000723.4 | NP_000714.3 | 1, 2 | 300 | ||
NM_199247.2 | NP_954855.1 | 1, 2 | 300 | |||
NM_199248.2 | NP_954856.1 | 1, 2 | 300 | |||
XM_005257645.2 | XP_005257702.1 | 1, 2 | 235 | |||
XM_005257646.2 | XP_005257703.1 | 1, 2 | 235 | |||
XM_017025027.1 | XP_016880516.1 | 1, 2 | 213 | |||
XM_017025030.1 | XP_016880519.1 | 1, 2 | 235 | |||
GenBank mRNA | AB054985.1 | - | 1, 2 | 253 | ||
AK289729.1 | - | 1, 2 | 266 | |||
AK297643.1 | - | 1, 2 | 299 | |||
AK312385.1 | - | 1, 2 | 299 | |||
AY393857.1 | - | 1, 2 | 92 | |||
BC037311.1 | - | 1, 2 | 237 | 5262182 | ||
BP217857.1 | - | 1, 2 | 300 | |||
M76560.1 | - | 1, 2 | 92 | |||
M92301.1 | - | 1, 2 | 241 | |||
M92302.1 | - | 1, 2 | 241 | |||
M92303.1 | - | 1, 2 | 241 |