Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037598.1 | - | 3 | 313 | ||
AK132675.1 | - | 5 | 500 | |||
AK157083.1 | - | 4 | 374 | |||
BC052907.1 | - | 4 | 379 | |||
BC098499.1 | - | 4 | 367 | 30074608 | ||
BY750146.1 | - | 4 | 365 | |||
U06923.1 | - | 4 | 370 | |||
U09351.1 | - | 4 | 824 |