Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011435.3 | NP_035565.1 | 2 | 1707 | ||
GenBank mRNA | AK075570.1 | - | 2 | 1562 | ||
AK145194.1 | - | 1566 | ||||
BC010975.1 | - | 2 | 1525 | 3983917 | ||
U38261.1 | - | 2 | 1708 |