Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011479.3 | NP_035609.1 | 4 | 551 | ||
GenBank mRNA | AK044917.1 | - | 4 | 552 | ||
AK057644.1 | - | 4 | 587 | |||
AK089019.1 | - | 4 | 545 | |||
AK159346.1 | - | 4 | 552 | |||
BC003227.1 | - | 4 | 584 | 3483158 | ||
U27455.1 | - | 4 | 519 | |||
X95642.1 | - | 4 | 522 |