Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164071.1 | NP_001157543.1 | 3 | 363 | ||
NM_001164072.1 | NP_001157544.1 | 2 | 158 | |||
NM_011529.2 | NP_035659.1 | 2 | 154 | |||
GenBank mRNA | AK031559.1 | - | 2 | 120 | ||
AK038910.1 | - | 2 | 93 | |||
AK088817.1 | - | 2 | 104 | |||
AK147727.1 | - | 3 | 309 | |||
AK165420.1 | - | 2 | 114 | |||
AK165994.1 | - | 2 | 89 | |||
BB660164.1 | - | 2 | 159 | |||
BC026148.1 | - | 2 | 139 | 3492937 | ||
BC033282.1 | - | 2 | 90 | |||
BC052685.1 | - | 2 | 141 | |||
U51907.1 | - | 2 | 96 | |||
U59864.1 | - | 2 | 167 |