Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253862.1 | NP_001240791.1 | 4, 5 | 440 | ||
NM_011544.3 | NP_035674.2 | 4, 5 | 497 | |||
GenBank mRNA | AK134037.1 | - | 4, 5 | 441 | ||
AK143246.1 | - | 4, 5 | 500 | |||
AK169800.1 | - | 4, 5 | 526 | |||
BY739497.1 | - | 4, 5 | 498 | |||
M97635.1 | - | 4, 5 | 468 | |||
X64840.1 | - | 4, 5 | 297 |