Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000829.3 | NP_000820.3 | 11 | 1769 | ||
NM_001077243.2 | NP_001070711.2 | 11 | 1769 | |||
NR_046356.1 | 10 | 1637 | ||||
XM_005271518.3 | XP_005271575.1 | 11 | 1813 | |||
XM_006718823.1 | XP_006718886.1 | 11 | 1443 | |||
XM_011542775.2 | XP_011541077.1 | 11 | 1813 | |||
XM_011542776.2 | XP_011541078.1 | 9 | 3347 | |||
XM_011542777.2 | XP_011541079.1 | 8 | 3217 | |||
XM_017017609.1 | XP_016873098.1 | 11 | 1813 | |||
XM_017017610.1 | XP_016873099.1 | 11 | 1813 | |||
XM_017017611.1 | XP_016873100.1 | 8 | 1454 | |||
GenBank mRNA | AB209092.1 | - | 10 | 1607 | ||
BC114549.1 | - | 10 | 1322 | |||
U16129.1 | - | 10 | 1323 |