Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001094.4 | NP_001085.2 | 1 | 470 | ||
GenBank mRNA | AK313043.1 | - | 1 | 439 | ||
BC075042.2 | - | 1 | 237 | 30915358 | ||
BC075043.2 | - | 1 | 237 | 30915574 | ||
U50352.1 | - | 1 | 196 | |||
U53212.1 | - | 1 | 323 | |||
U57352.1 | - | 1 | 470 |