Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161722.1 | NP_001155194.1 | 9 | 1600 | ||
NM_001161723.1 | NP_001155195.1 | 9 | 1594 | |||
NM_011549.3 | NP_035679.3 | 8 | 1541 | |||
GenBank mRNA | AF079095.1 | - | 8 | 1522 | ||
AK078755.1 | - | 9 | 1536 | |||
AK146016.1 | - | 8 | 1566 | |||
AK154971.1 | - | 9 | 1598 | |||
AK212830.1 | - | 1 | 184 | |||
BC064789.1 | - | 8 | 1541 | 30429545 | ||
U36393.1 | - | 1 | 325 |