Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 3 | 435 | ||
NM_001318157.1 | NP_001305086.1 | 3 | 753 | |||
NM_001318158.1 | NP_001305087.1 | 3 | 435 | |||
NR_134513.1 | 3 | 435 | ||||
NR_134514.1 | 3 | 435 | ||||
NR_134515.1 | 3 | 435 | ||||
XM_005251453.3 | XP_005251510.1 | 4 | 535 | |||
XM_006716765.3 | XP_006716828.1 | 1 | 45 | |||
XM_011517866.2 | XP_011516168.1 | 3 | 345 | |||
XM_017014700.1 | XP_016870189.1 | 3 | 636 | |||
XM_017014701.1 | XP_016870190.1 | 3 | 430 | |||
XM_017014702.1 | XP_016870191.1 | 3 | 314 | |||
XM_017014703.1 | XP_016870192.1 | 3 | 218 | |||
XM_017014704.1 | XP_016870193.1 | 1 | 45 | |||
XM_017014706.1 | XP_016870195.1 | 3 | 315 | |||
GenBank mRNA | AB006190.1 | - | 3 | 272 | ||
AK294028.1 | - | 3 | 219 | |||
AK297860.1 | - | 3 | 340 | |||
AK297885.1 | - | 3 | 569 | |||
AK301949.1 | - | 3 | 398 | |||
AK310456.1 | - | 1 | 147 | |||
BC119672.1 | - | 3 | 267 | |||
BC119673.1 | - | 3 | 267 | |||
DC306826.1 | - | 3 | 435 |
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 3 | 307 | ||
XM_006712884.3 | XP_006712947.1 | 3 | 731 | |||
XM_006712887.3 | XP_006712950.1 | 2 | 303 | |||
XM_017005467.1 | XP_016860956.1 | 2 | 303 | |||
XM_017005468.1 | XP_016860957.1 | 2 | 211 |