Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011594.3 | NP_035724.2 | 5 | 3230 | ||
GenBank mRNA | AK032818.1 | - | 5 | 3144 | ||
AK081522.1 | - | 5 | 3330 | |||
BC021538.1 | - | 1 | 216 | |||
BC024962.1 | - | 1 | 503 | |||
BC029809.1 | - | 1 | 155 | |||
BC034375.1 | - | 1 | 155 | |||
BC035468.1 | - | 1 | 330 | |||
BC037235.1 | - | 1 | 155 | |||
BC049126.1 | - | 5 | 3216 | 6821553 | ||
BC053932.1 | - | 1 | 328 |