Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277903.1 | NP_001264832.1 | 11 | 630 | ||
NM_001277904.1 | NP_001264833.1 | 10 | 594 | |||
NM_011618.2 | NP_035748.1 | 11 | 627 | |||
GenBank mRNA | AF020946.1 | - | 11 | 614 | ||
AJ131711.1 | - | 10 | 575 | |||
AK003317.1 | - | 3 | 209 | |||
AK160226.1 | - | 11 | 628 | |||
BC138871.1 | - | 11 | 607 | 8861893 | ||
BC141142.1 | - | 11 | 610 | 9055754 | ||
BC145449.1 | - | 10 | 574 | 9054007 | ||
BC145450.1 | - | 10 | 574 | 9054008 | ||
BC145451.1 | - | 11 | 610 | 9054009 | ||
U92883.1 | - | 11 | 611 | |||
U92884.1 | - | 10 | 578 |