Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011634.3 | NP_035764.2 | 15 | 1452 | ||
GenBank mRNA | AK012786.1 | - | 15 | 1434 | ||
AK012948.1 | - | 15 | 1451 | |||
BC006929.1 | - | 14 | 3177 | |||
BC017374.1 | - | 15 | 1441 | 3990371 | ||
U77844.1 | - | 15 | 1457 |