Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077591.1 | NP_001071059.1 | 9 | 1482 | ||
NM_011651.2 | NP_035781.2 | 9 | 1509 | |||
GenBank mRNA | AF025310.2 | - | 9 | 1489 | ||
BC118618.1 | - | 9 | 1461 | 40107271 | ||
BC121827.1 | - | 9 | 1461 | 40107270 |