Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011671.5 | NP_035801.3 | 5, 6 | 907 | ||
GenBank mRNA | AF111998.1 | - | 3, 4 | 610 | ||
AF111999.1 | - | 3, 4 | 604 | |||
AK035298.1 | - | 902 | ||||
AK054060.1 | - | 908 | ||||
AK134116.1 | - | 5, 6 | 908 | |||
AK142687.1 | - | 5, 6 | 908 | |||
AK165245.1 | - | 5, 6 | 908 | |||
BC012697.1 | - | 5, 6 | 906 | 4205625 | ||
BC012967.1 | - | 4, 5 | 808 | 2811421 | ||
U69135.1 | - | 5, 6 | 835 | |||
U94593.1 | - | 5, 6 | 828 |