Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011672.4 | NP_035802.3 | 2 | 239 | ||
GenBank mRNA | AK010461.1 | - | 2 | 208 | ||
AK088823.1 | - | 2 | 208 | |||
AK147138.1 | - | 2 | 222 | |||
AK169062.1 | - | 2 | 221 | |||
BC006630.1 | - | 2 | 196 | 3708320 | ||
CJ085208.1 | - | 2 | 239 | |||
U64445.1 | - | 2 | 224 |