Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011673.3 | NP_035803.1 | 4 | 456 | ||
GenBank mRNA | AK138848.1 | - | 4 | 472 | ||
BC050828.1 | - | 4 | 456 | 6512920 | ||
D89866.1 | - | 4 | 547 |