Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011703.4 | NP_035833.2 | 4 | 521 | ||
GenBank mRNA | AF266282.1 | - | 4 | 403 | ||
AK052465.1 | - | 4 | 522 |
Set Membership: |
G-protein coupled receptor |