Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011709.5 | NP_035839.3 | 2 | 158 | ||
GenBank mRNA | AK142655.1 | - | 159 | |||
AK164735.1 | - | 2 | 159 | |||
BC132280.1 | - | 2 | 156 | 40130557 | ||
BC132282.1 | - | 2 | 156 | 40130559 | ||
V00856.1 | - | 2 | 146 |