Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 22 | 3162 | ||
GenBank mRNA | AK083423.1 | - | 11 | 1568 | ||
AK143582.1 | - | 22 | 3097 | |||
AK149964.1 | - | 22 | 3120 | |||
AK163201.1 | - | 5 | 808 | |||
AK164245.1 | - | 13 | 1937 | |||
AK169372.1 | - | 23 | 3141 | |||
AK169933.1 | - | 22 | 3132 | |||
AK171271.1 | - | 22 | 3132 | |||
AK171633.1 | - | 22 | 3115 | |||
AK181180.1 | - | 1 | 80 | |||
AK188202.1 | - | 1 | 94 | |||
AK193942.1 | - | 1 | 268 | |||
BC011341.1 | - | 21 | 2939 | 3661165 | ||
BC066180.1 | - | 22 | 3072 | |||
BC066865.1 | - | 22 | 3072 | |||
L00681.1 | - | 22 | 3126 |