Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293757.1 | NP_001280686.1 | 4 | 549 | ||
NM_001293758.1 | NP_001280687.1 | 3 | 382 | |||
NM_001293759.1 | NP_001280688.1 | 4 | 619 | |||
NM_011722.4 | NP_035852.1 | 3 | 436 | |||
GenBank mRNA | AF124788.1 | - | 3 | 167 | ||
AF190796.1 | - | 3 | 177 | |||
AK003618.1 | - | 3 | 176 | |||
AK088312.1 | - | 3 | 183 | |||
BC029249.1 | - | 3 | 177 | 4486579 | ||
BY079663.1 | - | 3 | 236 | |||
CV560309.1 | - | 3 | 333 | |||
DV042569.1 | - | 3 | 391 | |||
DV062065.1 | - | 4 | 341 |