Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011782.2 | NP_035912.2 | 1 | 965 | ||
GenBank mRNA | AF140673.1 | - | 1 | 124 | ||
BC138619.1 | - | 1 | 149 | 8861639 | ||
BC138620.1 | - | 1 | 149 | 8861640 |