Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011804.2 | NP_035934.1 | 1, 2 | 367 | ||
GenBank mRNA | AF084524.1 | - | 1, 2 | 367 | ||
AK005388.1 | - | 1, 2 | 388 | |||
AK010947.1 | - | 1, 2 | 384 | |||
AK085844.1 | - | 1, 2 | 179 | |||
AK144101.1 | - | 1, 2 | 386 | |||
AK190952.1 | - | 1, 2 | 76 | |||
AK191906.1 | - | 1, 2 | 76 | |||
AK193253.1 | - | 1, 2 | 82 | |||
AK194678.1 | - | 1, 2 | 77 | |||
AK203453.1 | - | 1, 2 | 76 | |||
AK204882.1 | - | 1, 2 | 76 | |||
AK219109.1 | - | 1, 2 | 71 | |||
BC027426.1 | - | 1, 2 | 367 | 3670649 |
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK214038.1 | - | 336 |