Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079844.2 | NP_001073313.1 | 2 | 833 | ||
NM_011821.3 | NP_035951.1 | 2 | 833 | |||
GenBank mRNA | AF105268.1 | - | 3 | 556 | ||
AK032866.1 | - | 2 | 254 | |||
AK132531.1 | - | 2 | 570 | |||
AK135442.1 | - | 2 | 815 | |||
AK149312.1 | - | 2 | 600 | |||
AK164668.1 | - | 2 | 835 | |||
BC023448.1 | - | 2 | 247 | 5008374 |