Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF074018.1 | - | 2, 3 | 151 | ||
AK087774.1 | - | 3, 4 | 219 | |||
AY196361.1 | - | 2, 3 | 151 | |||
AY196362.1 | - | 2, 3 | 151 | |||
AY329484.1 | - | 2, 3 | 151 | |||
AY329485.1 | - | 2, 3 | 151 | |||
AY329486.1 | - | 2, 3 | 151 | |||
AY329487.1 | - | 2, 3 | 151 | |||
AY329488.1 | - | 2, 3 | 151 | |||
AY329489.1 | - | 2, 3 | 151 | |||
AY329490.1 | - | 2, 3 | 151 | |||
AY329491.1 | - | 2, 3 | 151 | |||
BC053384.1 | - | 3, 4 | 218 | 30049556 |